Please login to your online account to display your discounted pricing

Sino Biological™ CX3CL1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CX3CL1 (NP_002987.1) extracellular domain (Met 1-Arg 339) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10636H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10636H08H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 35kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin
Common Name Chemokine (C-X3-C motif) ligand 1
Gene Symbol CX3CL1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Chemokines & Receptors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 6376

Chemokine (C-X3-C Motif) Ligand 1 (CX3CL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to compete with biotinylated recombinant human CX3CL1-His for binding to immobilized recombinant mouse CD4-his (Mfr. No. 50134-M08) in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.