Please login to your online account to display your discounted pricing

Sino Biological™ CTSS Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the pro form of human CTSS (NP_004070.3) (Met 1-Ile 331) fused to a carboxy-terminal polyhistidine tag

Manufacturer: life technologies  LS10487H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10487H08H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 37kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Proteases
Endotoxin Concentration <1 EU/μg
Gene Alias MGC3886, CTSS
Common Name Cathepsin S
Gene Symbol CTSS
Protein Tag His Tag
Validated Application Assay Standard
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen

Cathepsin S (CTSS), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave a fluorogenic peptide substrate, (7-methoxycoumarin-4-yl) acetyl-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys (2, 4-dinitrophenyl)-NH2 (R∓D Systems Cat. No. ES002); cleavage of ES002 can be measured using excitation and emission wavelengths at 320nM and 405nM, respectively, the specific activity is >300pmoles/min./μg
  • Formulation: 12.5mM MES, 0.075M NaCl, 10% glycerol, pH 6.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.