Please login to your online account to display your discounted pricing

Sino Biological™ CTSL1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the pro form of human CTSL1 (NP_001903.1) (Met 1-Val 333) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10486H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10486H08H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 37.3kDa
Purity ≥85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Proteases|Glycoproteins
Endotoxin Concentration <1 EU/μg
Gene Alias CTSL1, CATL, CTSL, FLJ31037, MEP
Common Name Cathepsin L1
Gene Symbol CTSL1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Cathepsin L1 (CTSL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD74 at 5μg/mL (100μL/well) can bind biotinylated human CTSL1 with a linear range of 3.2-400ng/mL
  • Formulation: Lyophilized in 50mM NaAc, 0.1M NaCl, pH 5.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.