missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CTSL1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the pro form of human CTSL1 (NP_001903.1) (Met 1-Val 333) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10486H08H5

 View more versions of this product

Catalog No. 10486H08H5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CTSL1 Recombinant Human Protein, His Tag; 5ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Cathepsin L1 (CTSL1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD74 at 5μg/mL (100μL/well) can bind biotinylated human CTSL1 with a linear range of 3.2-400ng/mL
  • Formulation: Lyophilized in 50mM NaAc, 0.1M NaCl, pH 5.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
37.3kDa
5μg
Proteases|Glycoproteins
CTSL1, CATL, CTSL, FLJ31037, MEP
CTSL1
Other Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Cathepsin L1
Assay Standard
Mammalian