missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CTSB Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the pre pro form of human CTSB (NP_001899.1) fused to a C-terminal polyhistidine tag

$232.00 - $232.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias APPS, CPSB, CTSB
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10483H08H5 View Documents Life Technologies
LS10483H08H5
5μg Each for $232.00
 
Description

Description

Cathepsin B (CTSB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate Z-LR-AMC (R&D Systems Cat. No. ES008), the specific activity is >2500pmoles/min./μg
  • Formulation: Lyophilized in 25mM Tris, 100mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
APPS, CPSB, CTSB

For Research Use Only. Not for use in diagnostic procedures.