Please login to your online account to display your discounted pricing

Sino Biological™ CTSB Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the pre pro form of human CTSB (NP_001899.1) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10483H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10483H08H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 37.2kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Proteases
Endotoxin Concentration <1 EU/μg
Gene Alias APPS, CPSB, CTSB
Common Name Cathepsin B
Gene Symbol CTSB
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Cathepsin B (CTSB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate Z-LR-AMC (R&D Systems Cat. No. ES008), the specific activity is >2500pmoles/min./μg
  • Formulation: Lyophilized in 25mM Tris, 100mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.