Please login to your online account to display your discounted pricing

Sino Biological™ CTSA Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human cathepsin A isoform b (Met 1-Tyr 480) (NP_001121167.1) fused to a N-terminal signal peptide and a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10482H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10482H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 53kDa
Purification Method PAGE
Purity ≥85% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Full Length/Recombinant
Protein Subtype Proteases|Peptidase
Endotoxin Concentration <1 EU/μg
Gene Alias CTSA, RP3-337O18.1, GLB2, GSL, NGBE, PPCA, PPGB
Common Name Cathepsin A
Gene Symbol CTSA
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Proteases, Peptidases & Inhibitors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 5476

Cathepsin A (CTSA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.