missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CTLA4 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CTLA4 (NP_033973.2) (Met 1-Phe 162) fused to a polyhistidine tag at the C-terminus

$65.00 - $510.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CH29-120A16.2, Cd152, Ctla-4, Ly-56, Ctla4
View More Specs

Includes: Five vials of protein in suspension buffer

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50503M08H25 View Documents Life Technologies
LS50503M08H25
5 x 5μg Each for $164.00
 
50503M08H5 View Documents Life Technologies
LS50503M08H5
5μg Each for $65.00
 
50503M08H50 View Documents Life Technologies
LS50503M08H50
50μg Each for $250.00
 
50503M08HL View Documents Life Technologies
LS50503M08H250
5 x 50μg Each for $510.00
 
Description

Description

Cytotoxic T-Lymphocyte-Associated 4 (CTLA4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized recombinant mouse CTLA4-His (Mfr. No. 50503-M08H) at 10μg/mL (100μL/well) can mouse CD80-Fc (Mfr. No. 50446-M02H) with a linear range of 0.78-62.5ng/mL; 2. Immobilized recombinant mouse CTLA4-His (Mfr. No. 50503-M08H) at 10μg/mL (100μL/well) can human B7-1-Fc (Mfr. No. 10698-H03H) with a linear range of 0.78-125ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CH29-120A16.2, Cd152, Ctla-4, Ly-56, Ctla4
PAGE
Lyophilized
12477

For Research Use Only. Not for use in diagnostic procedures.