missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CTLA4 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human CTLA4 (NP_005205.2) (Met 1-Phe 162) fused to a polyhistidine tag at the C-terminus

$198.00 - $198.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CTLA4, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, ICOS, IDDM12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11159H08H50 View Documents Life Technologies
LS11159H08H50
50μg Each for $198.00
 
Description

Description

Cytotoxic T-lymphocyte-associated protein 4 (CTLA4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human CTLA4 at 10μg/mL (100μL/well) can bind human CD80-Fc, the EC50 of human CD80-Fc is 14ng/mL; 2. Immobilized human CTLA4 at 10μg/mL (100μL/well) can bind human CD86-His-Fc, the EC50 of human CD86-His-Fc is 53ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
CTLA4, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, ICOS, IDDM12

For Research Use Only. Not for use in diagnostic procedures.