Please login to your online account to display your discounted pricing

Sino Biological™ CSTA Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CSTA (NP_003801.1) (Thr 95-Gly 281) fused to a polyhistidinetag at the N-terminus

Manufacturer: life technologies  LS10479H07E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10479H07E5

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 1475
Endotoxin Concentration <1 EU/μg
Structural Form Full Length/Recombinant
Gene Alias STF1, STFA
Expression System E. coli
Purity ≥90% by SDS-PAGE
Molecular Weight 22.6kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Proteases, Peptidases & Inhibitors
Termination N-Terminal
Common Name Cystatin A
Purification Method PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Symbol CSTA
Quantity 5μg

Cystatin-A (CSTA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 1mM EDTA, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.