missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CSF3R (CD114) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 621) of human CSF3R precursor (NP_000751.1) precursor fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10218H08H250

 View more versions of this product

Catalog No. 10218H08HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CSF3R (CD114) Recombinant Human Protein, His Tag; 5 x 50ug; Source: Human; Purity: greater than 85% by SDS-PAGE

Colony stimulating factor 3 receptor (CSF3R/CD114), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its ability to inhibit the CSF3 induced proliferation of NFS60 mouse myeloid cells, the ED50 for this effect is typically 20-80ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1441
>85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Cytokine Receptors
CD114, GCSFR, CSF3R
CSF3R
Cytokines & Receptors
His Tag
Lyophilized
68kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Colony stimulating factor 3 receptor (granulocyte)
Assay Standard
Mammalian