Please login to your online account to display your discounted pricing

Sino Biological™ CSF3R (CD114) Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 621) of human CSF3R precursor (NP_000751.1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10218H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10218H02H50

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Gene ID (Entrez) 1441
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Protein Tag IgG1 Fc Tag
Quantity 50μg
Gene Alias CD114, GCSFR, CSF3R
Molecular Weight 93.3kDa
Purity ≥95% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Symbol CSF3R
Protein Subtype Cytokine Receptors
Validated Application Assay Standard
Protein Family Cytokines & Receptors/CD Proteins
Expression System Mammalian
Common Name Colony stimulating factor 3 receptor (granulocyte)

Colony stimulating factor 3 receptor (CSF3R/CD114), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard or in other research applications.

  • Activity: Measured by its ability to inhibit the GCSF-induced proliferation of NFS-60 mouse myeloid cells, the ED50 for this effect is typically 1-4ng/mL in the presence of 0.125ng/mL of recombinant human GCSF
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.