missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CSF2RB, Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 443) of human CSF2RB (NP_000386.1) fused to a polyhistidine-tag at the C-terminus

$209.00 - $209.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias CSF2RB, CD131, CDw131, IL3RB, IL5RB
Gene ID (Entrez) 1439
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10516H08H25 View Documents Life Technologies
LS10516H08H25
5 x 5μg Each for $209.00
 
Description

Description

Colony Stimulating Factor 2 Receptor, Beta, Low-Affinity (CSF2RB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human CD131 at 10μg/mL (100μL/well) can bind biotinylated human EPOR/Fc with a linear range of 0.16-4μg/mL; 2. Immobilized human CD131 at 10μg/mL (100μL/well) can bind human GM-CSF with a linear range of 0.032-4μg/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
1439
PAGE
CSF2RB, CD131, CDw131, IL3RB, IL5RB

For Research Use Only. Not for use in diagnostic procedures.