missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CSF2 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human CSF2 (NP_000749.2) (Ala 18-Glu 144) fused to a polyhistidine tag at the N-terminus

$96.50 - $96.50

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CSF2, GMCSF, mgC131935, mgC138897
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10015H07H5 View Documents Life Technologies
LS10015H07H5
5μg Each for $96.50
 
Description

Description

Colony stimulating factor 2 (granulocyte-macrophage) (CSF2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF-1 human erythroleukemic cells; the ED50 for this effect is typically 0.1-0.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Lyophilized
CSF2, GMCSF, mgC131935, mgC138897

For Research Use Only. Not for use in diagnostic procedures.