Please login to your online account to display your discounted pricing

Sino Biological™ CSF2 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human CSF2 (NP_000749.2) (Ala 18-Glu 144) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10015H07H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10015H07H5

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Recombinant/Inactive
Validated Application Assay Standard
Format Lyophilized
Protein Subtype Colony Stimulating Factors
Quantity 5μg
Common Name Colony stimulating factor 2 (granulocyte-macrophage)
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥85% by SDS-PAGE
Molecular Weight 16.9kDa
Protein Family Cytokines & Receptors
Expression System Mammalian
Gene Alias CSF2, GMCSF, mgC131935, mgC138897
Protein Tag His Tag
Endotoxin Concentration <1 EU/μg
Gene Symbol CSF2

Colony stimulating factor 2 (granulocyte-macrophage) (CSF2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured in a cell proliferation assay using TF-1 human erythroleukemic cells; the ED50 for this effect is typically 0.1-0.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.