missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CSF1R (MCSFR) Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu512) of human CSF1R precursor (NP_005202.2) fused to the a C-terminal polyhistidine tag

$230.00 - $800.00

Specifications

Purity >98% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias M-CSF R, CSF1R, FMS, CSFR, FIM2, C-FMS, CD115
Gene ID (Entrez) 1436
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10161H08H50 View Documents Life Technologies
LS10161H08H50
50μg Each for $230.00
 
10161H08HL View Documents Life Technologies
LS10161H08H250
5 x 50μg Each for $800.00
 
Description

Description

Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF1R at 10μg/mL can bind biotinylated human CSF1, the ED50 of biotinylated human CSF1 is 8ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

>98% by SDS-PAGE
Lyophilized
1436
<1 EU/μg
M-CSF R, CSF1R, FMS, CSFR, FIM2, C-FMS, CD115

For Research Use Only. Not for use in diagnostic procedures.