Please login to your online account to display your discounted pricing

Sino Biological™ CSF1R (MCSFR) Recombinant Human Protein (without Catalytic Activity Domain), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu512) of human CSF1R precursor (NP_005202.2) fused to the a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10161H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10161H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 56kDa
Quantity 5 x 5μg
Storage Requirements Store at -20 to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Cytokine Receptors
Endotoxin Concentration <1 EU/μg
Gene Alias M-CSF R, CSF1R, FMS, CSFR, FIM2, C-FMS, CD115
Gene Symbol CSF1R
Validated Application Assay Standard
Protein Family Growth Factors & Receptors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 1436
Purity >98% by SDS-PAGE
Common Name Colony stimulating factor 1 receptor
Protein Tag His Tag
Expression System Mammalian

Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF1R at 10μg/mL can bind biotinylated human CSF1, the ED50 of biotinylated human CSF1 is 8ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.