Please login to your online account to display your discounted pricing

Sino Biological™ CSF1R (MCSFR) Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu512) of human CSF1R precursor (NP_005202.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$69.50 - $772.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias M-CSF R, CSF1R, FMS, CSFR, FIM2, C-FMS, CD115
Gene ID (Entrez) 1436
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10161H03H5

DFS Item
 
life technologies
LS10161H03H5
5μg Each for $69.50

10161H03H25

DFS Item
 
life technologies
LS10161H03H25
5 x 5μg Each for $182.00

10161H03H50

DFS Item
 
life technologies
LS10161H03H50
50μg Each for $221.00

10161H03HL

DFS Item
 
life technologies
LS10161H03H250
5 x 50μg Each for $772.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias M-CSF R, CSF1R, FMS, CSFR, FIM2, C-FMS, CD115
Gene ID (Entrez) 1436

Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF1R at 1μg/well can bind biotinylated human CSF-1-His, the ED50 of biotinylated human CSF1-His is 8ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.