Please login to your online account to display your discounted pricing

Sino Biological™ CSF1R (MCSFR) Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu512) of human CSF1R precursor (NP_005202.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10161H03H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10161H03H25

  • / Each
 

Description & Specifications

Specifications

Gene Alias M-CSF R, CSF1R, FMS, CSFR, FIM2, C-FMS, CD115
Storage Requirements Store at -20 to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene ID (Entrez) 1436
Validated Application Assay Standard
Gene Symbol CSF1R
Endotoxin Concentration <1 EU/μg
Quantity 5 x 5μg
Protein Subtype Cytokine Receptors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Common Name Colony stimulating factor 1 receptor
Structural Form Truncated/Recombinant/Cell Surface Protein
Expression System Mammalian
Protein Family Growth Factors & Receptors
Molecular Weight 82.4kDa
Protein Tag His Tag/IgG1 Fc Tag
Purity ≥85% by SDS-PAGE

Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF1R at 1μg/well can bind biotinylated human CSF-1-His, the ED50 of biotinylated human CSF1-His is 8ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.