Please login to your online account to display your discounted pricing

Sino Biological™ CSF1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the N-terminal fragment (Met 1-Asn 190) of human CSF1 (P09603-1) fused to a polyhistidine tag at the C-terminus

$146.00 - $716.00

Specifications

Purity >98% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-195M16.2, MCSF, mgC31930
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11792H08H5

DFS Item
 
life technologies
LS11792H08H5
5μg Each for $146.00

11792H08H50

DFS Item
 
life technologies
LS11792H08H50
50μg Each for $716.00
Description & Specifications

Specifications

Purity >98% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-195M16.2, MCSF, mgC31930

Macrophage Colony Stimulating Factor (CSF1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF1R (100μL/well of a 10μg/mL solution) can bind biotinylated human CSF1; the EC50 of biotinylated human CSF1 is 8ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.