missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CRP Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding human CRP (NP_000558.2) (Met 1-Pro 224) fused to a polyhistidine tag at the C-terminus

$117.60 - $326.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Gene Alias RP11-419N10.4, mgC149895, mgC88244, PTX1
Gene ID (Entrez) 1401
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11250H08H25 Life Technologies
LS11250H08H25
5 x 5μg Each for $117.60
11LH08HL Life Technologies
LS11250H08H250
5 x 50μg Each for $326.00
Specifications

Description

C-Reactive Protein Pentraxin-Related (CRP) recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human CD32a (Mfr. No.10374-H08B) in functional ELISA
  • Formulation: Lyophilized in 20mM Tris, 0.2M NaCl, 5mM CaCl2, pH 8.0
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
1401
PAGE
RP11-419N10.4, mgC149895, mgC88244, PTX1