missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CRP Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding human CRP (NP_000558.2) (Met 1-Pro 224) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS11250H08H250

 View more versions of this product

Catalog No. 11LH08HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CRP Recombinant Human Protein, His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

C-Reactive Protein Pentraxin-Related (CRP) recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human CD32a (Mfr. No.10374-H08B) in functional ELISA
  • Formulation: Lyophilized in 20mM Tris, 0.2M NaCl, 5mM CaCl2, pH 8.0
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1401
PAGE
5 x 50μg
C-Terminal
RP11-419N10.4, mgC149895, mgC88244, PTX1
CRP
Other Proteins
His Tag
Lyophilized
24.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
C-reactive protein, pentraxin-related
Assay Standard
Mammalian