missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CNTNAP2 Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CNTNAP2 (NP_001004357.2) (Met 1-Ser 1262) fused to a C-terminal polyhistidine tag

$237.00 - $237.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CNTNAP2, 5430425M22Rik, Caspr2, Mkiaa0868
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50649M08H25 Life Technologies
LS50649M08H25
5 x 5μg Each for $237.00
Specifications

Description

Contactin Associated Protein-like 2 (CNTNAP2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of C6 Rat brain glial cells; mouse CNTNAP2 immobilized at 0.8μg/mL, 100μL/well will mediate >30% C6 cell adhesion
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CNTNAP2, 5430425M22Rik, Caspr2, Mkiaa0868
PAGE
Lyophilized
66797