Please login to your online account to display your discounted pricing

Sino Biological™ CNTF Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CNTF (NP_000605.1) (Ala 2-Met 200) fused to a polyhistide tag at the N-terminus

$317.00 - $453.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias HCNTF, CNTF
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11841H07E25

DFS Item
 
life technologies
LS11841H07E25
5 x 5μg Each for $317.00

11841H07E50

DFS Item
 
life technologies
LS11841H07E50
50μg Each for $453.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias HCNTF, CNTF

Ciliary Neurotrophic Factor (CNTF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human CTNFR at 10μg/mL (100μL/well) can bind human CNTF with a linear ranger of 1.28-160ng/mL; 2. Immobilized Rat CTNFR at 10μg/mL (100μL/well) can bind human CNTF with a linear range of 1.28-160ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.