missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CNDP1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human CNDP1 (NP_116038.4) (Ser 27-His 508) fused to a polyhistidine tag at the C-terminus

$1,040.00 - $3,590.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CNDP1, CN1, CPGL2, HsT2308, mgC10825, mgC102737, mgC142072
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10077H08H50 Life Technologies
LS10077H08H50
50μg Each for $1,040.00
10077H08HL Life Technologies
LS10077H08H250
5 x 50μg Each for $3,590.00
Specifications

Description

Carnosine Dipeptidase 1 (CNDP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
CNDP1, CN1, CPGL2, HsT2308, mgC10825, mgC102737, mgC142072
PAGE
Lyophilized
84735