missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CLU Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CLU isoform long (NP_001822.2) (Met 1-Glu 501) fused to a C-terminal polyhistidine tag

$97.75 - $211.05

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AAG4, APOJ, CLI, KUB1, mgC24903, SGP-2, SGP2, SP-40, TRPM-2, TRPM2
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11297H08H25 Life Technologies
LS11297H08H25
5 x 5μg Each for $211.05
11297H08H5 Life Technologies
LS11297H08H5
5μg Each for $97.75
Specifications

Description

Clusterin (CLU), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
AAG4, APOJ, CLI, KUB1, mgC24903, SGP-2, SGP2, SP-40, TRPM-2, TRPM2
PAGE
Lyophilized
1191