Please login to your online account to display your discounted pricing

Sino Biological™ CLMP (ASAM) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of the human ASAM precursor (NP_079045.1) (Met 1-Met 233) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10794H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10794H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 25.8kDa
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Gene Alias ASAM, ACAM, CLMP, FLJ22415
Common Name CXADR-like membrane protein
Protein Tag His Tag
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Gene Symbol CLMP
Validated Application Assay Standard
Format Lyophilized

CXADR-like membrane protein (ASAM), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of the HUVEC human umbilical vein endothelial cell line; when 4 x 104 cells/well are added to human ASAM coated plates (30μg/mL, 100μL/well), approximately 25-50 % will adhere after one hour at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.