missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CLEC12A (MICL) Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CLEC12A (EAW96132.1) extracellular domain (His 75-Ala 275) fused to a polyhistidine tag at the N-terminus

$1,166.00 - $1,166.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CLL-1, CLL1, DCAL-2, mgC70602, MICL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11896H07H50 View Documents Invitrogen
LS11896H07H50
50μg Each for $1,166.00
 
Description

Description

C-type Lectin domain family 12, member A (CLEC12A), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 7.4
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
<1 EU/μg
CLL-1, CLL1, DCAL-2, mgC70602, MICL
PAGE
Lyophilized
160364

For Research Use Only. Not for use in diagnostic procedures.