missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CHIT1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CHIT1 (NP_003456.1) (Met 1-Asn 466) precursor fused to a polyhistidine tag at the C-terminus

$586.00 - $586.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias CHI3, CHIT, FLJ00314, mgC125322
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11223H08H25 Life Technologies
LS11223H08H25
5 x 5μg Each for $586.00
Specifications

Description

Chitinase 1 (Chitotriosidase) (CHIT1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 12.5mM Tris, 75mM NaCl, 50% Glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CHI3, CHIT, FLJ00314, mgC125322
PAGE
Frozen
1118