missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CHIT1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CHIT1 (NP_003456.1) (Met 1-Asn 466) precursor fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS11223H08H25

 View more versions of this product

Catalog No. 11223H08H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CHIT1 Recombinant Human Protein, His Tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Chitinase 1 (Chitotriosidase) (CHIT1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 12.5mM Tris, 75mM NaCl, 50% Glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1118
PAGE
5 x 5μg
Full Length/Recombinant
CHI3, CHIT, FLJ00314, mgC125322
CHIT1
Cell Structure/Organelle Proteins
His Tag
Frozen
50.8kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Chitinase 1 (Chitotriosidase)
Assay Standard
Mammalian