missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CHI3L1 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CHI3L1 (Q61362) (Met 1-Ala 381) fused to a C-terminal polyhistidine tag

$207.00 - $364.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AW208766, Brp39, Gp39
View More Specs

Includes: Five vials of protein in suspension buffer

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50929M08H25 View Documents Invitrogen
LS50929M08H25
5 x 5μg Each for $207.00
 
50929M08H50 View Documents Invitrogen
LS50929M08H50
50μg Each for $364.00
 
Description

Description

Chitinase 3-like 1 (CHI3L1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
<1 EU/μg
AW208766, Brp39, Gp39
PAGE
Lyophilized
12654

For Research Use Only. Not for use in diagnostic procedures.