Please login to your online account to display your discounted pricing

Sino Biological™ CHI3L1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CHI3L1 (NP_001267.2) (Met 1-Thr 383) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11227H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11227H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 42kDa
Protein Family Other Proteins
Endotoxin Concentration <1 EU/μg
Format Frozen
Expression System Mammalian
Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Quantity 5 x 5μg
Purity ≥95% by SDS-PAGE
Common Name Chitinase 3-like 1 (cartilage glycoprotein-39)
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Symbol CHI3L1
Validated Application Assay Standard
Gene Alias ASRT7, DKFZp686N19119, FLJ38139, GP39, HC-gp39, HCGP-3P, YKL40, YYL-40
Protein Tag His Tag
Protein Subtype Other Proteins

Chitinase 3-Like 1 (Cartilage Glycoprotein-39) (CHI3L1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of Fadu human squamous cell carcinoma cells (ATCC: HTB43); when cells are added to recombinant human CHI3L1 coated plates (1.5μg/mL, 100μL/well), approximately 65-75% will adhere specifically after 30 minutes at 37°C
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.