Please login to your online account to display your discounted pricing

Sino Biological™ CFD (Adipsin) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human CFD (P00746) (Ile 26-Ala 253) fused to a C-terminal polyhistidine tag and a signal peptide at the N-terminus

Manufacturer: life technologies  LS10160H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10160H08H50

  • / Each
 

Description & Specifications

Specifications

Quantity 50μg
Purification Method PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name Complement factor D (adipsin)
Structural Form Full Length/Recombinant
Expression System Mammalian
Format Lyophilized
Purity ≥90% by SDS-PAGE
Validated Application Assay Standard
Gene Alias ADIPSIN, ADN, DF, PFD
Endotoxin Concentration <1 EU/μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Protein Tag His Tag
Gene ID (Entrez) 1675
Termination C-Terminal
Protein Family Other Proteins
Molecular Weight 25.8kDa
Gene Symbol CFD

Complement Factor D (CFD), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.