Please login to your online account to display your discounted pricing

Sino Biological™ CES2E (CES5) Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CES2E (NP_766347.1) (Met 1-His 556) fused to the polyhistidine tag at the C-terminus

$241.00 - $4,185.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 9030624L02Rik, CES5
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50620M08H25

DFS Item
 
life technologies
LS50620M08H25
5 x 5μg Each for $700.00

50620M08H5

DFS Item
 
life technologies
LS50620M08H5
5μg Each for $241.00

50620M08H50

DFS Item
 
life technologies
LS50620M08H50
50μg Each for $1,172.00

50620M08HL

DFS Item
 
life technologies
LS50620M08H250
5 x 50μg Each for $4,185.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 9030624L02Rik, CES5

Carboxylesterase 2E (CES2E), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to hydrolyze pnitrophenylacetate; the specific activity is >50nM/min./μg
  • Formulation: Lyophilized in 20mM Tris,150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.