missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CEACAM8 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CEACAM8 (NP_001807.2) without the pro peptide (Met 1-Ser319) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen LS11729H08H50

 View more versions of this product

Catalog No. 11729H08H50



Description

Description

Carcinoembryonic Antigen-Related Cell Adhesion Molecule 8 (CEACAM8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CEACAM6-his (Mfr. No. 10823-H08H) at 10μg/mL (100μL/well) can bind human CEACAM8; the EC50 value is 0.17μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Glycoproteins|Intercellular Adhesion Molecule
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
32.8kDa
50μg
<1 EU/μg
Carcinoembryonic antigen-related cell adhesion molecule 8
His Tag
CD Proteins
Assay Standard
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
CD66b, CD67, CGM6, NCA-95, CEACAM8
CEACAM8
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.