missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CEACAM6 (CD66C) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CEACAM6 (NP_002474.3) (Met 1-Gly 320) fused to a polyhistidine tag at the C-terminus

$95.75 - $1,320.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CEACAM6, CD66c, CEAL, NCA
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10823H08H25 Life Technologies
LS10823H08H25
5 x 5μg Each for $212.00
10823H08H5 Life Technologies
LS10823H08H5
5μg Each for $95.75
10823H08HL Life Technologies
LS10823H08H250
5 x 50μg Each for $1,320.00
Specifications

Description

Carcinoembryonic antigen-related cell adhesion molecule 6 (nonspecific cross reacting antigen) (CEACAM6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA; 2. Immobilized human CEACAM6-His (Mfr. No. 10823-H08H) at 10μg/mL (100μL/well) can bind biotinylated human CEACAM8-His, the EC50 of biotinylated human CEACAM8-His is 0.17μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
CEACAM6, CD66c, CEAL, NCA