missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CEACAM-5 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human CEACAM5 (NP_004354.2) (Met 1-Ala 685) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS11077H08H50

 View more versions of this product

Catalog No. 11077H08H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; CEACAM-5 Recombinant Human Protein, His Tag; 50ug; Source: Human; Purity: greater than or equal to 95% by SDS-PAGE

Carcinoembryonic Antigen-Related Cell Adhesion Molecule 5 Protein (CEACAM5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1048
PAGE
50μg
C-Terminal
<1 EU/μg
Carcinoembryonic Antigen-Related Cell Adhesion Molecule 5 Protein
Assay Standard
Mammalian
Lyophilized
72.8kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
CD66e, CEA, DKFZp781M2392, CEACAM5
CEACAM5
CD Proteins
His Tag