Please login to your online account to display your discounted pricing

Sino Biological™ CEACAM-5 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human CEACAM5 (NP_004354.2) (Met 1-Ala 685) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11077H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11077H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 72.8kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥95% by SDS-PAGE
Gene ID (Entrez) 1048
Purification Method PAGE
Gene Symbol CEACAM5
Quantity 5μg
Common Name Carcinoembryonic Antigen-Related Cell Adhesion Molecule 5 Protein
Structural Form Full Length/Recombinant
Protein Tag His Tag
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
Termination C-Terminal
Protein Family CD Proteins
Protein Subtype Glycoproteins
Expression System Mammalian
Gene Alias CD66e, CEA, DKFZp781M2392, CEACAM5
Format Lyophilized

Carcinoembryonic Antigen-Related Cell Adhesion Molecule 5 Protein (CEACAM5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.