missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CDH17 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CDH17 (Q12864-1) extracellular domain (Met 1-Met 787) fused to a polyhistidine tag at the C-terminus

$424.00 - $424.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CDH16, FLJ26931, HPT-1, HPT1, mgC138218, mgC142024
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11360H08H50 View Documents Life Technologies
LS11360H08H50
50μg Each for $424.00
 
Description

Description

Cadherin 17, LI cadherin (liver-intestine) (CDH17), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of MCF-7 human breast adenocarcinoma cells; when 5 x 104 cells/well are added to recombinant human Cadherin 17 coated plates (0.8μg/mL with 100μL/well), approximately >50% will adhere after 1 hour at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Lyophilized
CDH16, FLJ26931, HPT-1, HPT1, mgC138218, mgC142024

For Research Use Only. Not for use in diagnostic procedures.