Please login to your online account to display your discounted pricing

Sino Biological™ CDC42 Recombinant Human Protein, GST Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form (Met 1-Cys 188) of human CDC42 isoform 2 (P60953-2) fused to the GST tag at the N-terminus

$88.25 - $554.00

Specifications

Purity >85% by SDS-PAGE
Format Lyophilized
Gene Alias RP1-224A6.5, CDC42Hs, G25K
Gene ID (Entrez) 998
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10062H09E25

DFS Item
 
life technologies
LS10062H09E25
5 x 5μg Each for $153.00

10062H09E5

DFS Item
 
life technologies
LS10062H09E5
5μg Each for $88.25

10062H09E50

DFS Item
 
life technologies
LS10062H09E50
50μg Each for $226.00

10062H09EL

DFS Item
 
life technologies
LS10062H09E250
5 x 50μg Each for $554.00
Description & Specifications

Specifications

Purity >85% by SDS-PAGE
Format Lyophilized
Gene Alias RP1-224A6.5, CDC42Hs, G25K
Gene ID (Entrez) 998

Cell division cycle 42 (CDC42), GTP binding protein of 25kDa, recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 20mM Tris buffer, 150mM NaCl, 0.5mM reduced Glutathione, pH 8.0; contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.