Please login to your online account to display your discounted pricing

Sino Biological™ CDC42 Recombinant Human Protein, GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form (Met 1-Cys 188) of human CDC42 isoform 2 (P60953-2) fused to the GST tag at the N-terminus

Manufacturer: life technologies  LS10062H09E250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10062H09EL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 48.1kDa
Quantity 5 x 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias RP1-224A6.5, CDC42Hs, G25K
Common Name Cell division cycle 42
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Purity >85% by SDS-PAGE
Structural Form Recombinant
Protein Subtype GTPases
Gene Symbol CDC42
Protein Tag GST Tag
Validated Application Assay Standard
Protein Family Other Proteins
Gene ID (Entrez) 998

Cell division cycle 42 (CDC42), GTP binding protein of 25kDa, recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 20mM Tris buffer, 150mM NaCl, 0.5mM reduced Glutathione, pH 8.0; contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.