missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CDC37 Recombinant Human Protein, GST Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the full length of human CDC37 (NP_008996.1) (Met 1-Val 378) fused to the GST tag at the C-terminus

$143.00 - $350.00

Specifications

Purity >85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias P50CDC37
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10647H09B25 Life Technologies
LS10647H09B25
5 x 5μg Each for $350.00
10647H09B5 Life Technologies
LS10647H09B5
5μg Each for $143.00
Specifications

Description

Cell Division Cycle 37 homolog (S. cerevisiae) (CDC37), recombinant human protein is supplied in frozen format. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 0.5mM PMSF, 10% glycerol, 1mM GSH, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>85% by SDS-PAGE
<1 EU/μg
P50CDC37
PAGE
Frozen
11140