Please login to your online account to display your discounted pricing

Sino Biological™ CDC37 Recombinant Human Protein, GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the full length of human CDC37 (NP_008996.1) (Met 1-Val 378) fused to the GST tag at the C-terminus

Manufacturer: life technologies  LS10647H09B5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10647H09B5

  • / Each
 

Description & Specifications

Specifications

Quantity 5μg
Validated Application Assay Standard
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 70.7kDa
Expression System Insect
Purity >85% by SDS-PAGE
Purification Method PAGE
Gene ID (Entrez) 11140
Structural Form Full Length/Recombinant
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Format Frozen
Common Name Cell Division Cycle 37 Homolog (S. cerevisiae)
Gene Alias P50CDC37
Endotoxin Concentration <1 EU/μg
Protein Tag GST Tag
Gene Symbol CDC37

Cell Division Cycle 37 homolog (S. cerevisiae) (CDC37), recombinant human protein is supplied in frozen format. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 0.5mM PMSF, 10% glycerol, 1mM GSH, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.