Please login to your online account to display your discounted pricing

Sino Biological™ CDC37 Recombinant Human Protein, GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the full length of human CDC37 (NP_008996.1) (Met 1-Val 378) fused to the GST tag at the C-terminus

Manufacturer: life technologies  LS10647H09B25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10647H09B25

  • / Each
 

Description & Specifications

Specifications

Format Frozen
Gene Symbol CDC37
Quantity 5 x 5μg
Purity >85% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Expression System Insect
Endotoxin Concentration <1 EU/μg
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Molecular Weight 70.7kDa
Gene ID (Entrez) 11140
Purification Method PAGE
Common Name Cell Division Cycle 37 Homolog (S. cerevisiae)
Protein Tag GST Tag
Structural Form Full Length/Recombinant
Gene Alias P50CDC37
Protein Family Other Proteins

Cell Division Cycle 37 homolog (S. cerevisiae) (CDC37), recombinant human protein is supplied in frozen format. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 0.5mM PMSF, 10% glycerol, 1mM GSH, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.