Please login to your online account to display your discounted pricing

Sino Biological™ CD96 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CD96 isoform 2 (P40200-2) extracellular domain (Met 1-Met 503) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11202H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11202H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 55kDa
Quantity 5 x 5μg
Gene ID (Entrez) 10225
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Protein Family CD Proteins
Gene Symbol CD96
Expression System Mammalian
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Common Name CD96 Molecule
Protein Tag His Tag
Gene Alias CD96
Validated Application Assay Standard
Purity ≥85% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Structural Form Truncated/Recombinant
Protein Subtype Other Proteins
Termination C-Terminal
Purification Method PAGE

Cluster of Differentiation 96 (CD96), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: 1. Measured by its ability to bind recombinant mouse PVR in a functional ELISA; 2. Measured by its ability to bind recombinant Human CD155 in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.