missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD8A Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Tyr 196) of mouse CD8A (NP_001074579.1) fused to a polyhistidine tag at the C-terminus

$343.00 - $1,742.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD8A, BB154331, Ly-2, Ly-35, Ly-B, Lyt-2
Gene ID (Entrez) 12525
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50389M08H25 View Documents Life Technologies
LS50389M08H25
5 x 5μg Each for $343.00
 
50389M08HL View Documents Life Technologies
LS50389M08H250
5 x 50μg Each for $1,742.00
 
Description

Description

Cluster of Differentiation 8A (CD8A), recombinant ferret protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: 1. Measured by its ability to bind biotinylated recombinant human B2M in a functional ELISA; 2. Measured by its ability to bind biotinylated recombinant human FCGRT and B2M in a functional ELISA; 3. Measured by its ability to bind biotinylated recombinant human LCK in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
12525
<1 EU/μg
CD8A, BB154331, Ly-2, Ly-35, Ly-B, Lyt-2

For Research Use Only. Not for use in diagnostic procedures.