missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD86 (B7-2) Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 245) of mouse CD86 precursor (NP_062261.3) fused to a polyhistidine tag at the C-terminus

$393.00 - $393.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD86, B7, B7-2, B7.2, B70, CLS1, CD28l2, ETC-1, Ly-58, Ly58, MB7, MB7-2, TS/A-2
Gene ID (Entrez) 12524
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50068M08H50 View Documents Invitrogen
LS50068M08H50
50μg Each for $393.00
 
Description

Description

Cluster of Differentiation 86 (CD86/B7-2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse CD86 at 20μg/mL (100μL/well) can bind mouse CD28 with a linear range of 6.4-800ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
12524
<1 EU/μg
CD86, B7, B7-2, B7.2, B70, CLS1, CD28l2, ETC-1, Ly-58, Ly58, MB7, MB7-2, TS/A-2

For Research Use Only. Not for use in diagnostic procedures.