missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD86 (B7-2) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 239) of human CD86 (NP_008820.2) fused to a polyhistidine tag at the C-terminus

$90.50 - $716.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias B7-2, B70, CD28LG2, LAB72, mgC34413
Gene ID (Entrez) 942
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10699H08H25 Life Technologies
LS10699H08H25
5 x 5μg Each for $173.00
10699H08H50 Life Technologies
LS10699H08H50
50μg Each for $253.00
10699H08H5 Life Technologies
LS10699H08H5
5μg Each for $90.50
10699H08HL Life Technologies
LS10699H08H250
5 x 50μg Each for $716.00
Specifications

Description

Cluster of Differentiation 86 (CD86/B7-2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD86 at 20μg/mL (100μL/well) can bind human CD28 with a linear range of 32-800ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
942
<1 EU/μg
B7-2, B70, CD28LG2, LAB72, mgC34413