Please login to your online account to display your discounted pricing

Sino Biological™ CD86 (B7-2) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 239) of human CD86 (NP_008820.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10699H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10699H08H5

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Quantity 5μg
Gene ID (Entrez) 942
Protein Family CD Proteins
Molecular Weight 26.2kDa
Format Lyophilized
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Structural Form Truncated/Recombinant/Cell Surface Protein
Validated Application Assay Standard
Purity ≥90% by SDS-PAGE
Common Name CD86 molecule
Protein Tag His Tag
Gene Symbol CD86
Gene Alias B7-2, B70, CD28LG2, LAB72, mgC34413
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Expression System Mammalian

Cluster of Differentiation 86 (CD86/B7-2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD86 at 20μg/mL (100μL/well) can bind human CD28 with a linear range of 32-800ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.