missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CD84 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 225) of human CD84 (NP_003865.1) fused to a polyhistidine tag at the C-terminus

$121.00 - $1,382.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias DKFZp781E2378, LY9B, SLAMF5, hCD84, mCD84.
Gene ID (Entrez) 8832
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10100H08H25 Life Technologies
LS10100H08H25
5 x 5μg Each for $246.00
10100H08H5 Life Technologies
LS10100H08H5
5μg Each for $121.00
10100H08H50 Life Technologies
LS10100H08H50
50μg Each for $417.00
10100H08HL Life Technologies
LS10100H08H250
5 x 50μg Each for $1,382.00
Specifications

Description

Cluster of Differentiation 84 (CD84), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
8832
<1 EU/μg
DKFZp781E2378, LY9B, SLAMF5, hCD84, mCD84.